Skip to Content
Merck
All Photos(1)

Key Documents

73489

Sigma-Aldrich

L-α-Neopentylglycine

≥98.0% (TLC)

Synonym(s):

γ-Methyl-L-leucine, (S)-2-Amino-4,4-dimethylpentanoic acid, L-γ-Methylleucine

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$481.00

$481.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$481.00

About This Item

Empirical Formula (Hill Notation):
C7H15NO2
CAS Number:
Molecular Weight:
145.20
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

$481.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

Assay

≥98.0% (TLC)

form

solid

reaction suitability

reaction type: solution phase peptide synthesis

application(s)

peptide synthesis

SMILES string

[H][C@](N)(CC(C)(C)C)C(O)=O

InChI

1S/C7H15NO2/c1-7(2,3)4-5(8)6(9)10/h5H,4,8H2,1-3H3,(H,9,10)/t5-/m0/s1

InChI key

LPBSHGLDBQBSPI-YFKPBYRVSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J L Fauchère et al.
International journal of peptide and protein research, 18(3), 249-255 (1981-09-01)
The modified Strecker synthesis of Patel & Worsley was combined with an enzymatic deamidation in the final step, to produce the title compound. The amino acid is a member of the family of the "fat" amino-acids with high lipophilicity and
Xuejing Yu et al.
The FEBS journal, 281(1), 391-400 (2013-11-12)
Branched-chain amino acid aminotransferase (BCAT) plays a key role in the biosynthesis of hydrophobic amino acids (such as leucine, isoleucine and valine), and its substrate spectrum has not been fully explored or exploited owing to the inescapable restrictions of previous

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service