Skip to Content
Merck
All Photos(1)

Key Documents

731722

Sigma-Aldrich

Copper(II) tert-butylacetoacetate

97%

Synonym(s):

Bis(tert-butylacetoacetato)copper(II)

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$623.00

$623.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
10 G
$623.00

About This Item

Empirical Formula (Hill Notation):
C16H26CuO6
CAS Number:
Molecular Weight:
377.92
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

$623.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Assay

97%

form

solid

reaction suitability

core: copper
reagent type: catalyst

mp

105-110 °C

SMILES string

CC(=O)\C=C(/O[Cu]O\C(OC(C)(C)C)=C/C(C)=O)OC(C)(C)C

InChI

1S/2C8H14O3.Cu/c2*1-6(9)5-7(10)11-8(2,3)4;/h2*5,10H,1-4H3;/q;;+2/p-2/b2*7-5+;

InChI key

CYHFRTGEBHOGBO-XXXYRMOLSA-L

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service