Skip to Content
Merck
All Photos(1)

Key Documents

729779

Sigma-Aldrich

4-Bromo-3-methoxyphenol

97%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$40.70

$40.70

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$40.70

About This Item

Empirical Formula (Hill Notation):
C7H7BrO2
CAS Number:
Molecular Weight:
203.03
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$40.70

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Assay

97%

form

solid

mp

77-82 °C

functional group

bromo

SMILES string

COc1cc(O)ccc1Br

InChI

1S/C7H7BrO2/c1-10-7-4-5(9)2-3-6(7)8/h2-4,9H,1H3

InChI key

UYDZUCNMZXCLJI-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service