Skip to Content
Merck
All Photos(1)

Key Documents

728438

Sigma-Aldrich

4-(3-Chloropropyl)morpholine hydrochloride

97%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$92.80

$92.80


Estimated to ship onApril 19, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$92.80

About This Item

Empirical Formula (Hill Notation):
C7H14ClNO · HCl
CAS Number:
Molecular Weight:
200.11
Beilstein:
3684151
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$92.80


Estimated to ship onApril 19, 2025


Request a Bulk Order

Quality Level

Assay

≥96.5% (GC)
96.5-103.5% (AT)
97%

form

crystals

functional group

chloro
ether

SMILES string

Cl.ClCCCN1CCOCC1

InChI

1S/C7H14ClNO.ClH/c8-2-1-3-9-4-6-10-7-5-9;/h1-7H2;1H

InChI key

PQECODMSWJOUAT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service