Skip to Content
Merck
All Photos(1)

Key Documents

727997

Sigma-Aldrich

Fmoc-Cys(4-methoxytrityl)-OH

≥98% (HPLC), for peptide synthesis

Synonym(s):

(R)-2-(9H-Fluoren-9-ylmethoxycarbonylamino)-3-[(4-methoxyphenyl)diphenylmethylsulfanyl]propionic acid, Fmoc-Cys(Mmt)-OH, Nα-(9-Fluorenylmethyloxycarbonyl)-S-4-methoxytrityl-L-cysteine

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$305.00

$305.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$305.00

About This Item

Empirical Formula (Hill Notation):
C38H33NO5S
CAS Number:
Molecular Weight:
615.74
Beilstein:
8524562
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

$305.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Product Name

Fmoc-Cys(4-methoxytrityl)-OH, ≥98% (HPLC)

Assay

≥98% (HPLC)

form

lumps

optical activity

[α]/D +14.0±1.0°, c = 1 in THF

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

COc1ccc(cc1)C(SC[C@H](NC(=O)OCC2c3ccccc3-c4ccccc24)C(O)=O)(c5ccccc5)c6ccccc6

InChI

1S/C38H33NO5S/c1-43-29-22-20-28(21-23-29)38(26-12-4-2-5-13-26,27-14-6-3-7-15-27)45-25-35(36(40)41)39-37(42)44-24-34-32-18-10-8-16-30(32)31-17-9-11-19-33(31)34/h2-23,34-35H,24-25H2,1H3,(H,39,42)(H,40,41)/t35-/m0/s1

InChI key

LOBUWFUSGOYXQX-DHUJRADRSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elyse T Williams et al.
Angewandte Chemie (International ed. in English), 57(36), 11640-11643 (2018-07-07)
We report a new method herein coined SP-CLipPA (solid-phase cysteine lipidation of a peptide or amino acid) for the synthesis of mono-S-lipidated peptides. This technique utilizes thiol-ene chemistry for conjugation of a vinyl ester to a free thiol of a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service