Skip to Content
Merck
All Photos(1)

Key Documents

339016

Sigma-Aldrich

Tetrafluorophthalic anhydride

97%

Synonym(s):

3,4,5,6-Tetrafluorophthalic anhydride, 4,5,6,7-Tetrafluoro-1,3-isobenzofurandione

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$197.00

$197.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$197.00

About This Item

Empirical Formula (Hill Notation):
C8F4O3
CAS Number:
Molecular Weight:
220.08
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$197.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

97%

mp

94-96 °C (lit.)

SMILES string

Fc1c(F)c(F)c2C(=O)OC(=O)c2c1F

InChI

1S/C8F4O3/c9-3-1-2(8(14)15-7(1)13)4(10)6(12)5(3)11

InChI key

BJDDKZDZTHIIJB-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Fluorocarbon polymers exhibit increased thermal stability, hydrophobicity, lipophobicity, and chemical resistance compared to hydrocarbon analogs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service