Skip to Content
Merck
All Photos(1)

Key Documents

726419

Sigma-Aldrich

2-(Chloromethyl)naphthalene

≥97.0%

Synonym(s):

β-Naphthylmethyl chloride, 2-Menaphthyl chloride, 2-Naphthylmethyl chloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$269.00

$269.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$269.00

About This Item

Empirical Formula (Hill Notation):
C11H9Cl
CAS Number:
Molecular Weight:
176.64
Beilstein:
742185
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$269.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥97.0% (GC)
≥97.0%
97.0-103.0% (T)

form

crystals

functional group

chloro

SMILES string

ClCc1ccc2ccccc2c1

InChI

1S/C11H9Cl/c12-8-9-5-6-10-3-1-2-4-11(10)7-9/h1-7H,8H2

InChI key

MPCHQYWZAVTABQ-UHFFFAOYSA-N

Application



  • Improving insulin sensitivity: Research highlights dihydroberberine′s capacity to enhance insulin sensitivity through AMPK activation, offering a promising therapeutic strategy for managing insulin resistance and type 2 diabetes (Lewis and Falk, 2022).


  • Anti-inflammatory effects: A study conducted on the anti-inflammatory properties of dihydroberberine found significant reductions in inflammatory markers, suggesting its potential as an anti-inflammatory agent in metabolic syndrome (Dian et al., 2022).


  • Enhancing bioavailability: Investigations into the oral bioavailability of dihydroberberine demonstrate improvements over its precursor berberine, highlighting its superior pharmacokinetic profile which could lead to more effective clinical applications (Murakami, Bodor, and Bodor, 2023).


  • Glucose uptake mechanisms: Studies on dihydroberberine have shown its effectiveness in facilitating glucose uptake, an essential function for metabolic health, through its interaction with cellular transport mechanisms (Moon et al., 2021).



Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service