Skip to Content
Merck
All Photos(1)

Key Documents

25170

Sigma-Aldrich

1-(Chloromethyl)naphthalene

technical, ≥97.0% (GC)

Synonym(s):

1-Naphthylmethyl chloride

Sign Into View Organizational & Contract Pricing

Select a Size

50 ML
$96.70

$96.70

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 ML
$96.70

About This Item

Linear Formula:
C10H7CH2Cl
CAS Number:
Molecular Weight:
176.64
Beilstein:
636885
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$96.70

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

technical

Assay

≥97.0% (GC)

form

solid

color

deep brown

refractive index

n20/D 1.635 (lit.)
n20/D 1.635

bp

167-169 °C/25 mmHg (lit.)

mp

32 °C (lit.)

density

1.18 g/mL at 25 °C (lit.)

functional group

chloro

SMILES string

ClCc1cccc2ccccc12

InChI

1S/C11H9Cl/c12-8-10-6-3-5-9-4-1-2-7-11(9)10/h1-7H,8H2

InChI key

XMWGTKZEDLCVIG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Kinetics and dissociation mechanism of 1-(chloromethyl)naphthalene following 266nm photoexcitation has been studied[1]. Intermolecular nucleophilic dearomatization of 1-chloromethyl naphthalene with various activated methylene compounds has been investigated[2].

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

269.6 °F - closed cup

Flash Point(C)

132 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Bo Peng et al.
Organic letters, 13(19), 5402-5405 (2011-09-14)
Palladium-catalyzed nucleophilic dearomatization of chloromethyl naphthalene derivatives to produce ortho- or para-substituted carbocycles in satisfactory to excellent yields has been developed. The unprecedented dearomatization reactions proceeded smoothly under mild conditions via η(3)-benzylpalladium intermediates.
Formation kinetics and spectra of aromatic radicals in solution.
Kelley DF, et al.
The Journal of Physical Chemistry, 87(11), 1842-1843 (1983)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service