Skip to Content
Merck
All Photos(1)

Key Documents

726206

Sigma-Aldrich

Bis(4-tert-butylphenyl)iodonium hexafluorophosphate

98%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$155.00
5 G
$599.00

$155.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$155.00
5 G
$599.00

About This Item

Empirical Formula (Hill Notation):
C20H26F6IP
CAS Number:
Molecular Weight:
538.29
Beilstein:
5721061
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

$155.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥97.5% (TLC)
98%

form

solid

SMILES string

F[P-](F)(F)(F)(F)F.CC(C)(C)c1ccc([I+]c2ccc(cc2)C(C)(C)C)cc1

InChI

1S/C20H26I.F6P/c1-19(2,3)15-7-11-17(12-8-15)21-18-13-9-16(10-14-18)20(4,5)6;1-7(2,3,4,5)6/h7-14H,1-6H3;/q+1;-1

InChI key

CJLLNCQWBHTSCB-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Man-Yu Duan et al.
Physical chemistry chemical physics : PCCP, 19(3), 2353-2358 (2017-01-06)
In this study, we demonstrate a novel method for fabricating polymer stabilized cholesteric liquid crystal (PSCLC) films with non-uniform pitch distribution by utilizing two kinds of photo-induced processes. Based on the large HTP temperature dependence of a chiral dopant, polymer
Shaohui Liu et al.
Polymers, 12(6) (2020-06-26)
In this article, different substituents (benzoyl, acetyl, styryl) are introduced onto the carbazole scaffold to obtain 8 novel carbazole derivatives. Interestingly, a benzoyl substituent, connected to a carbazole group, could form a benzophenone moiety, which composes a monocomponent Type II
Shaohui Liu et al.
Macromolecular rapid communications, 41(23), e2000460-e2000460 (2020-09-23)
In this study, a new generation of photoinitiator (PI) based on hybrid structures combining benzophenone and triphenylamine is proposed. Remarkably, these photoinitiators (noted monofunctional benzophenone-triphenylamine (MBP-TPA) and trifunctional benzophenone-triphenylamine (TBP-TPA)) are designed and developed for the photopolymerization under light-emitting diodes
Haifaa Mokbel et al.
Macromolecular rapid communications, 41(15), e2000289-e2000289 (2020-07-09)
Free radical polymerization upon near-infrared (NIR) light is still the subject of intense research efforts and remains a huge challenge particularly for long wavelengths (>1000 nm). In this study, a NIR sensitizer operating upon long wavelength (1064 nm) is proposed for an
Alvaro Iregui et al.
Polymers, 11(3) (2019-04-10)
In this work Poly ε-caprolactone (PCL)/ Diglycidyl ether of bisphenol A (DGEBA) blends were electrospun and the obtained mats were UV cured to achieve shape memory properties. In the majority of studies, when blends with different compositions are electrospun, the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service