Skip to Content
Merck
All Photos(1)

Key Documents

726192

Sigma-Aldrich

Bis(4-methylphenyl)iodonium hexafluorophosphate

98%

Synonym(s):

Bis(p-tolyl)iodonium hexafluorophosphate

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$304.00
25 G
$1,170.00

$304.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$304.00
25 G
$1,170.00

About This Item

Empirical Formula (Hill Notation):
C14H14F6IP
CAS Number:
Molecular Weight:
454.13
Beilstein:
3581445
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

$304.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥97.5% (TLC)
98%

form

solid

SMILES string

F[P-](F)(F)(F)(F)F.Cc1ccc([I+]c2ccc(C)cc2)cc1

InChI

1S/C14H14I.F6P/c1-11-3-7-13(8-4-11)15-14-9-5-12(2)6-10-14;1-7(2,3,4,5)6/h3-10H,1-2H3;/q+1;-1

InChI key

LHLVGWWCRPPKBC-UHFFFAOYSA-N

General description

Bis(4-methylphenyl)iodonium hexafluorophosphate is a white powder. It melts at 175-180 °C and shows an UV absorption peak at 267nm. 1

Application


  • Covalent Functionalisation of Few-Layer Black Phosphorus using Aryl Iodonium Salts and Comparison to Diazonium Modified Black Phosphorus: Explores the functionalization of black phosphorus using bis(4-methylphenyl)iodonium hexafluorophosphate and compares it to diazonium modification (M Van Druenen, 2019).

  • Iodonium sulfonates as high‐performance coinitiators and additives for CQ‐based systems: Toward aromatic amine‐free photoinitiating systems: Examines the effectiveness of iodonium sulfonates and bis(4-methylphenyl)iodonium hexafluorophosphate in CQ-based photoinitiating systems (J Kirschner et al., 2019).

  • Thiophene-substituted phenothiazine-based photosensitisers for radical and cationic photopolymerization reactions under visible laser beams (405 and 455 nm): Studies the use of thiophene-substituted phenothiazine in combination with bis(4-methylphenyl)iodonium hexafluorophosphate for effective photopolymerization (P Chao et al., 2016).

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service