Skip to Content
Merck
All Photos(1)

Key Documents

725625

Sigma-Aldrich

Caffeine-(trimethyl-d9)

99 atom % D, 98% (CP)

Synonym(s):

1,3,7-Trimethyl-d9-xanthine

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$618.00

$618.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

Select a Size

Change View
100 MG
$618.00

About This Item

Empirical Formula (Hill Notation):
C8D9HN4O2
Molecular Weight:
203.25
MDL number:
UNSPSC Code:
12352114
PubChem Substance ID:
NACRES:
NA.12

$618.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

isotopic purity

99 atom % D

Quality Level

Assay

98% (CP)

form

powder

technique(s)

mass spectrometry (MS): suitable

mp

234-236.5 °C (lit.)

mass shift

M+9

SMILES string

O=C(N(C([2H])([2H])[2H])C1=O)N(C2=C1N(C([2H])([2H])[2H])C=N2)C([2H])([2H])[2H]

InChI

1S/C8H10N4O2/c1-10-4-9-6-5(10)7(13)12(3)8(14)11(6)2/h4H,1-3H3/i1D3,2D3,3D3

InChI key

RYYVLZVUVIJVGH-GQALSZNTSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Catarina Rendeiro et al.
The British journal of nutrition, 116(12), 1999-2010 (2017-01-10)
Specific flavonoid-rich foods/beverages are reported to exert positive effects on vascular function; however, data relating to effects in the postprandial state are limited. The present study investigated the postprandial, time-dependent (0-7 h) impact of citrus flavanone intake on vascular function.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service