Skip to Content
Merck
All Photos(1)

Key Documents

724068

Sigma-Aldrich

1-Methylcyclopropanol

Synonym(s):

1-Methyl-1-cyclopropanol

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$197.00

$197.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
$197.00

About This Item

Empirical Formula (Hill Notation):
C4H8O
CAS Number:
Molecular Weight:
72.11
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$197.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

liquid

refractive index

n20/D 1.414

density

1.265 g/mL at 25 °C

functional group

hydroxyl

SMILES string

CC1(O)CC1

InChI

1S/C4H8O/c1-4(5)2-3-4/h5H,2-3H2,1H3

InChI key

NCTCZGRRDXIGIY-UHFFFAOYSA-N

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

77.0 °F

Flash Point(C)

25 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service