Skip to Content
Merck
All Photos(1)

Key Documents

723460

Sigma-Aldrich

2-Naphthyl (1R,4R,7R)-7-isopropyl-5-methylbicyclo[2.2.2]octa-2,5-diene-2-carboxylate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H24O2
CAS Number:
Molecular Weight:
332.44
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

reaction suitability

reaction type: click chemistry

mp

90-94 °C

SMILES string

CC(C)[C@H]1C[C@@H]2C=C([C@H]1C=C2C)C(=O)Oc3ccc4ccccc4c3

InChI

1S/C23H24O2/c1-14(2)20-12-18-13-22(21(20)10-15(18)3)23(24)25-19-9-8-16-6-4-5-7-17(16)11-19/h4-11,13-14,18,20-21H,12H2,1-3H3/t18-,20-,21+/m1/s1

InChI key

GVTUQEGQEVADFM-NRSPTQNISA-N

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service