Skip to Content
Merck
All Photos(1)

Key Documents

721948

Supelco

Methanol-d4

NMR reference standard, 99.8 atom % D, NMR tube size 5 mm × 8 in.

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3OD
Molecular Weight:
36.07
EC Number:
UNSPSC Code:
41116107

grade

NMR reference standard

isotopic purity

99.8 atom % D

Assay

95% (CP)

technique(s)

NMR: suitable

NMR tube size

5 mm × 8 in.

format

neat

mass shift

M+4

Looking for similar products? Visit Product Comparison Guide

Quantity

5 mm O.D. tube contains 0.700 mL.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexis Lavaud et al.
Phytochemistry, 109, 103-110 (2014-12-04)
Ten tocotrienol derivatives, i.e., amplexichromanols (1-10), were isolated from stem bark of Garcinia amplexicaulis Vieill. ex Pierre collected in Caledonia. The structures of the compounds 1-5 were determined to be chromanol derivatives substituted by a polyprenyl chain oxidized in terminal
Cheng-Kun Lin et al.
Organic & biomolecular chemistry, 13(7), 2100-2107 (2014-12-20)
A straightforward synthesis of novel, 2-heterocyclyl polyhydroxylated pyrrolidines is described. Stereocontrolled additions of nucleophiles to cyclic nitrones generated the corresponding 2,3-trans adducts, allowing the synthesis of the corresponding pyrrolidines via key intermediates bearing an alkyne and a nitrile oxide. Three
Jon Kapla et al.
Physical chemistry chemical physics : PCCP, 17(34), 22438-22447 (2015-08-08)
The disaccharide trehalose (TRH) strongly affects the physical properties of lipid bilayers. We investigate interactions between lipid membranes formed by 1,2-dimyristoyl-sn-glycero-3-phosphocholine (DMPC) and TRH using NMR spectroscopy and molecular dynamics (MD) computer simulations. We compare dipolar couplings derived from DMPC/TRH
Athina Dimopoulou et al.
Nucleosides, nucleotides & nucleic acids, 34(4), 289-308 (2015-03-17)
We report the synthesis of novel thiopurine pyranonucleosides. Direct coupling of silylated 6-mercaptopurine and 6-thioguanine with the appropriate pyranoses 1a-e via Vorbrüggen nucleosidation, gave the N-9 linked mercaptopurine 2a-e and thioguanine 4a-e nucleosides, while their N-7 substituted congeners 10a-e and
Joonki Kim et al.
Journal of agricultural and food chemistry, 63(31), 6979-6988 (2015-07-29)
We found that an extract of Lycoris chejuensis and its three isolated active components, narciclasine, 7-deoxynarciclasine, and 7-deoxy-trans-dihydronarciclasine, each significantly reduced the formation of amyloid-β peptides in HeLa cells transfected with an amyloid precursor protein carrying the Swedish mutation up

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service