Skip to Content
Merck
All Photos(2)

Key Documents

721689

Sigma-Aldrich

5-2,4-Cyclopentadien-1-yl)[(1,2,3-η)-1-phenyl-2-propenyl]-palladium

95%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$304.00

$304.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$304.00

About This Item

Linear Formula:
Pd(η5-C5H5)(η3-1-PhC3H4)
CAS Number:
Molecular Weight:
288.68
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

$304.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Assay

95%

form

solid

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Cross Couplings
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

mp

45-47 °C

storage temp.

2-8°C

SMILES string

[Pd].[CH]1[CH][CH][CH][CH]1.[CH2][CH][CH]c2ccccc2

InChI

1S/C9H9.C5H5.Pd/c1-2-6-9-7-4-3-5-8-9;1-2-4-5-3-1;/h2-8H,1H2;1-5H;

InChI key

STDKYMKMBSTDJK-UHFFFAOYSA-N

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Sol. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

nwg

Flash Point(F)

118.0 °F - closed cup

Flash Point(C)

47.8 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service