Skip to Content
Merck
All Photos(1)

Key Documents

721522

Sigma-Aldrich

2-(Aminomethyl)thiazole

97%

Synonym(s):

(2-Thiazolyl)methylamine

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$496.00

$496.00


Estimated to ship onMay 23, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$496.00

About This Item

Empirical Formula (Hill Notation):
C4H6N2S
CAS Number:
Molecular Weight:
114.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$496.00


Estimated to ship onMay 23, 2025


Request a Bulk Order

Assay

97%

form

liquid

refractive index

n20/D 1.569

density

1.204 g/mL at 25 °C

functional group

amine

SMILES string

NCc1nccs1

InChI

1S/C4H6N2S/c5-3-4-6-1-2-7-4/h1-2H,3,5H2

InChI key

JOZSYOPADROCMP-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

219.9 °F - closed cup

Flash Point(C)

104.4 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service