Skip to Content
Merck
All Photos(1)

Key Documents

710326

Sigma-Aldrich

(1,3-Dioxolan-2-ylmethyl)zinc bromide

0.6 M in THF

Sign Into View Organizational & Contract Pricing

Select a Size

50 ML
$627.00

$627.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 ML
$627.00

About This Item

Empirical Formula (Hill Notation):
C4H7BrO2Zn
Molecular Weight:
232.39
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

$627.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

liquid

reaction suitability

reaction type: C-C Bond Formation

concentration

0.6 M in THF

density

0.992 g/mL at 25 °C

functional group

ether

storage temp.

2-8°C

SMILES string

Br[Zn]CC1OCCO1

InChI

1S/C4H7O2.BrH.Zn/c1-4-5-2-3-6-4;;/h4H,1-3H2;1H;/q;;+1/p-1

InChI key

LOJRLQRGBKTHDB-UHFFFAOYSA-M

General description

(1,3-Dioxolan-2-ylmethyl)zinc bromide is an organozinc compound used as a reagent in Negishi cross-coupling reaction to prepare aryl or heteroaryl scaffolds via C-C bond formation.[1]

Application

(1,3-Dioxolan-2-ylmethyl)zinc bromide can be used as a reagent in the allylic alkylation reactions in the presence of iridium catalyst.[2]

Legal Information

Product of Rieke Metals, Inc.
Rieke is a registered trademark of Rieke Metals, Inc.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-22.0 °F

Flash Point(C)

-30 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of 8-C-substituted 2, 6-diaminopurine acyclic nucleoside phosphonates by Negishi cross-coupling
Sedlavcek O, et al.
Collection of Czechoslovak Chemical Communications, 10(26), 449-450 (2015)
Iridium-Catalyzed Enantioselective Allylic Alkylation with Functionalized Organozinc Bromides
Hamilton JY, et al.
Angewandte Chemie (International ed. in English), 54(26), 7644-7647 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service