Skip to Content
Merck
All Photos(1)

Key Documents

721492

Sigma-Aldrich

3-Aminobenzylamine

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H10N2
CAS Number:
Molecular Weight:
122.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

mp

41-45 °C

functional group

amine

SMILES string

NCc1cccc(N)c1

InChI

1S/C7H10N2/c8-5-6-2-1-3-7(9)4-6/h1-4H,5,8-9H2

InChI key

ZDBWYUOUYNQZBM-UHFFFAOYSA-N

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gonzalo Pérez-Mitta et al.
Journal of the American Chemical Society, 137(18), 6011-6017 (2015-04-17)
The ability to modulate the surface chemical characteristics of solid-state nanopores is of great interest as it provides the means to control the macroscopic response of nanofluidic devices. For instance, controlling surface charge and polarity of the pore walls is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service