Skip to Content
Merck
All Photos(1)

Key Documents

745731

Sigma-Aldrich

Dipropargylamine

97%

Synonym(s):

4-Azahepta-1,6-diyne, N,N-Di(prop-2-ynyl)amine, N-(2-Propyn-1-yl)-2-propyn-1-amine, N-(2-Propynyl)-2-propyn-1-amine, Di-2-propynylamine, NSC 80652

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$93.40

$93.40

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$93.40

About This Item

Empirical Formula (Hill Notation):
C6H7N
CAS Number:
Molecular Weight:
93.13
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$93.40

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.477

density

0.900 g/mL at 25 °C

functional group

amine

SMILES string

C#CCNCC#C

InChI

1S/C6H7N/c1-3-5-7-6-4-2/h1-2,7H,5-6H2

InChI key

RGSODMOUXWISAG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

115.0 °F - closed cup

Flash Point(C)

46.11 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kristen Bennett et al.
Applied and environmental microbiology, 82(8), 2270-2279 (2016-01-31)
Nitrosomonas europaea is an aerobic nitrifying bacterium that oxidizes ammonia (NH3) to nitrite (NO2 (-)) through the sequential activities of ammonia monooxygenase (AMO) and hydroxylamine dehydrogenase (HAO). Many alkynes are mechanism-based inactivators of AMO, and here we describe an activity-based

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service