Skip to Content
Merck
All Photos(1)

Key Documents

719129

Sigma-Aldrich

Plexcore® OC RG-1115 organic conductive ink

Synonym(s):

Plexcore® OC organic conductive ink

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352103

form

liquid

concentration

2.0%

resistivity

10-300 Ω-cm

work function

4.8 eV

pH

3.5

viscosity

8.7 cP(25 °C)

density

0.934 g/mL at 25 °C

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Warning

Do not freeze. Quality is compromised if ink goes through freeze-thaw cycle even once.

Physical properties

Composition: solids in water, 1,2-propanediol and isopropanol

Legal Information

Plexcore is a registered trademark of Plextronics, Inc.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

80.1 °F

Flash Point(C)

26.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service