Skip to Content
Merck
All Photos(1)

Key Documents

717703

Sigma-Aldrich

2,5-Bis(tributylstannyl)thiophene

97%

Synonym(s):

1,1′-(2,5-Thiophenediyl)bis[1,1,1-tributylstannane], 2,5-Bis(tri-n-butylstannyl)thiophene

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$108.00
5 G
$373.00

$108.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$108.00
5 G
$373.00

About This Item

Empirical Formula (Hill Notation):
C28H56SSn2
CAS Number:
Molecular Weight:
662.23
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

$108.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Assay

97%

form

liquid

density

1.201 g/mL at 25 °C

SMILES string

CCCC[Sn](CCCC)(CCCC)c1ccc(s1)[Sn](CCCC)(CCCC)CCCC

InChI

1S/C4H2S.6C4H9.2Sn/c1-2-4-5-3-1;6*1-3-4-2;;/h1-2H;6*1,3-4H2,2H3;;

InChI key

ITAZQNRIMIQTDI-UHFFFAOYSA-N

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhiwei Chu et al.
Sensors (Basel, Switzerland), 18(5) (2018-05-16)
An aggregation-caused quenching (ACQ)-active polymer (PF), an aggregation-induced emission (AIE)-active polymer (PFTPE) and an aggregation-enhanced emission (AEE)-active polymer (PTTPE) were synthesized by tetraphenylethane (TPE), fluorene and thiophene moieties. Polyurethane (PU) foams modified by PF, PFTPE and PTTPE, namely PU-PF, PU-PFTPE

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service