Skip to Content
Merck
All Photos(1)

Key Documents

717657

Sigma-Aldrich

2-Methoxy-1-naphthaleneboronic acid

Synonym(s):

(2-Methoxy-1-naphthalenyl)boronic acid, (2-Methoxy-1-naphthyl)boronic acid, 1-(2-Methoxynaphthyl)boronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H11BO3
CAS Number:
Molecular Weight:
202.01
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

solid

mp

125-130 °C

SMILES string

COc1ccc2ccccc2c1B(O)O

InChI

1S/C11H11BO3/c1-15-10-7-6-8-4-2-3-5-9(8)11(10)12(13)14/h2-7,13-14H,1H3

InChI key

NHVWTZOWDLOBBS-UHFFFAOYSA-N

Application

Reactant for:
  • Design of phosphorus C,P-ligands for environmentally benign cross-coupling reactions
  • Sterically hindered Suzuki-Miyaura coupling
  • Palladium-catalyzed cross-coupling reactions
  • Deboronation homocoupling reactions
  • Copper-catalyzed N-arylation of imidazoles
  • Stereoselective preparation of biaryl compounds via palladium-catalyzed asymmetric Suzuki-Miyaura coupling

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service