717169
2-Keto-3-methylbutyric acid-3-d sodium salt hydrate
98 atom % D, 98% (CP)
Synonym(s):
α-Keto-3-methyl-butyric acid-3-d sodium hydrate, α-Ketoisovaleric acid-3-d sodium salt hydrate, 2-Ketoisovaleric acid-3-d sodium salt hydrate, 3-Methyl-2-oxobutanoic acid-3-d sodium salt hydrate, Sodium α-Ketoisovalerate-3-d
About This Item
Recommended Products
isotopic purity
98 atom % D
Quality Level
Assay
98% (CP)
form
solid
technique(s)
bio NMR: suitable
mass shift
M+1
SMILES string
O.[Na+].[2H]C(C)(C)C(=O)C([O-])=O
InChI
1S/C5H8O3.Na.H2O/c1-3(2)4(6)5(7)8;;/h3H,1-2H3,(H,7,8);;1H2/q;+1;/p-1/i3D;;
InChI key
PGCNKYBBFKQNIR-KIGHCJKZSA-M
Packaging
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
We presents an informational article concerning biomolecular NMR and the use of Isotope Labeling Methods for Protein Dynamics Studies.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service