Skip to Content
Merck
All Photos(2)

Key Documents

615862

Sigma-Aldrich

L-Tryptophan-(indole-d5)

97 atom % D

Synonym(s):

(S)-2-Amino-3-(3-indoyl-d5)propionic acid, L-α-Amino-(3-indole-d5)propionic acid, Deuterated Trp, Deuterated tryptophan, Trp-d5

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$2,420.00

$2,420.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

Select a Size

Change View
500 MG
$2,420.00

About This Item

Empirical Formula (Hill Notation):
C11D5H7N2O2
CAS Number:
Molecular Weight:
209.26
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.12

$2,420.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

isotopic purity

97 atom % D

Quality Level

form

solid

optical activity

[α]20/D -30.5°, c = 1 in H2O

mp

280-285 °C (dec.) (lit.)

mass shift

M+5

SMILES string

[2H]c1[nH]c2c([2H])c([2H])c([2H])c([2H])c2c1C[C@H](N)C(O)=O

InChI

1S/C11H12N2O2/c12-9(11(14)15)5-7-6-13-10-4-2-1-3-8(7)10/h1-4,6,9,13H,5,12H2,(H,14,15)/t9-/m0/s1/i1D,2D,3D,4D,6D

InChI key

QIVBCDIJIAJPQS-HLTLGYGQSA-N

Related Categories

General description

L-Tryptophan-(indole-d5) is an isotopically labeled analogue of L-Tryptophan.

Application

L-Tryptophan-(indole-d5) is used
• in biophysical study for neutron scattering.
• as an internal standard for the investigation of anti-inflammatory activity of low molecular weight fractions.
• in structural elucidation of micronutrients and nutraceutical components presence in food stuffs using LC-MS/MS.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Valerie Laux et al.
European biophysics journal : EBJ, 37(6), 815-822 (2008-02-16)
We describe methods that have been developed within the ILL-EMBL Deuteration Laboratory for the production of maltose binding protein (MBP) that has been selectively labelled either with deuterated tryptophan or deuterated methionine (single labelling), or both (double labelling). MBP is
Gregory W Thomas et al.
Journal of immunoassay & immunochemistry, 37(1), 55-67 (2015-05-12)
The innate immune system is increasingly being recognized as a critical component in osteoarthritis (OA) pathophysiology. An ex vivo immunoassay utilizing human peripheral blood mononuclear cells (PBMC) was developed in order to assess the OA anti-inflammatory properties of the low
Sara Vitalini et al.
Molecules (Basel, Switzerland), 25(2) (2020-01-17)
Food plants contain hundreds of bioactive phytochemicals arising from different secondary metabolic pathways. Among these, the metabolic route of the amino acid Tryptophan yields a large number of plant natural products with chemically and pharmacologically diverse properties. We propose the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service