Skip to Content
Merck
All Photos(3)

Key Documents

715247

Sigma-Aldrich

Magnesium borohydride

greener alternative

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Mg(BH4)2
CAS Number:
Molecular Weight:
53.99
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23

Assay

95%

form

powder

reaction suitability

reagent type: reductant

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

SMILES string

[BH4-].[BH4-].[Mg++]

InChI

1S/2BH4.Mg/h2*1H4;/q2*-1;+2

InChI key

BAJRVYFABMRJEO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Magnesium borohydride (Mg(BH4)2) is a hydrogen storage material with a high gravimetric wt% of ~14.8 and a volumetric hydrogen density of 112 g/L. It has a relatively low hydrogen-release temperature and reversibility. Mg(BH4)2 can be used as an electrolytic material for rechargeable magnesium batteries. Its unsolvated form can be derived by solid state exchange reaction between MgCl2 and MBH4.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Used in hydrogen storage.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Skin Corr. 1B - Water-react 2

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High concentration magnesium borohydride/tetraglyme electrolyte for rechargeable magnesium batteries
Tuerxun F, et al.
Journal of Power Sources, 276, 255-261 (2015)
Magnesium borohydride: synthesis and crystal structure
Cerny R, et al.
Angewandte Chemie (International ed. in English), 46(30), 5765-5767 (2007)
Magnesium borohydride: synthesis and crystal structure.
Cerny R, et al.
Angewandte Chemie (International Edition in English), 119(30), 5867-5869 (2007)
Structure of unsolvated magnesium borohydride Mg (BH4) 2
Her JH, et al.
Acta Crystallographica Section B, Structural Science, 63(4), 561-568 (2007)
Magnesium borohydride as a hydrogen storage material: properties and dehydrogenation pathway of unsolvated Mg (BH 4) 2
Soloveichik GL, et al.
International Journal of Hydrogen Energy, 34(2), 916-928 (2009)

Articles

An article about metal borohydrides as hydrogen storage materials

Mechanochemical Effect of Severe Plastic Deformations: Metal Alloys, Hydrides and Molecular Solids

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service