Skip to Content
Merck
All Photos(2)

Key Documents

389986

Sigma-Aldrich

Calcium borohydride bis(tetrahydrofuran)

Synonym(s):

Calcium(+2) borohydride bis(tetrahydrofuran)

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$229.00

$229.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$229.00

About This Item

Linear Formula:
Ca(BH4)2· 2C4H8O
CAS Number:
Molecular Weight:
213.97
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

$229.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

powder

reaction suitability

reagent type: reductant

functional group

ether

SMILES string

[BH4-].[BH4-].[Ca++].C1CCOC1.C2CCOC2

InChI

1S/2C4H8O.2BH4.Ca/c2*1-2-4-5-3-1;;;/h2*1-4H2;2*1H4;/q;;2*-1;+2

InChI key

QNDQIIWFODWXIW-UHFFFAOYSA-N

Application

  • Reactant for preparation of calcium tetradecahydroundecaborate used for synthesizing other salts with boron hydride nido-borates
Calcium borohydride bis(tetrahydrofuran) can be used as a selective reducing agent to reduce α,β-unsaturated lactone derivatives (pterocidin) to corresponding diols without reducing the conjugated C-C double bond.[1]

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Skin Corr. 1B - Water-react 2

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Absolute configuration of pterocidin, a potent inhibitor of tumor cell invasion from a marine-derived Streptomyces
Igarashi Y, et al.
Tetrahedron Letters, 53(6), 654-656 (2012)

Articles

An article about metal borohydrides as hydrogen storage materials

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service