Skip to Content
Merck
All Photos(2)

Documents

714747

Sigma-Aldrich

Biphenyl-3,4′,5-tricarboxylic acid

greener alternative

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10O6
CAS Number:
Molecular Weight:
286.24
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:

Assay

96%

form

powder

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

>300 °C

greener alternative category

SMILES string

OC(=O)c1ccc(cc1)-c2cc(cc(c2)C(O)=O)C(O)=O

InChI

1S/C15H10O6/c16-13(17)9-3-1-8(2-4-9)10-5-11(14(18)19)7-12(6-10)15(20)21/h1-7H,(H,16,17)(H,18,19)(H,20,21)

InChI key

LQEZHWGJSWHXPJ-UHFFFAOYSA-N

General description

Biphenyl-3,4′,5-tricarboxylic acid (H3BPT) is a co-ordination polymer, which is majorly used as an unsymmetrically substituted linker. It forms self-assembly under the solvothermal condition to synthesize 3D lanthanide based metal organic frameworks (MOFs).
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Biphenyl-3,4′,5-tricarboxylic acid (H3BPT) may be used to prepare porous metal-organic frameworks (MOFs) for gas adsorption, photocatalytic activity and magnetic properties. H3BPT and (bis)imidazole linkers may be used to prepare coordination polymers (CPs) by hydrothermal process. CPs may be used in gas storage, catalysis, ion exchange etc.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Two (3, 6)-connected porous metal?organic frameworks based on linear trinuclear [Co 3 (COO) 6] and paddlewheel dinuclear [Cu 2 (COO) 4] SBUs: gas adsorption, photocatalytic behaviour, and magnetic properties.
Zhao J, et al.
Journal of Material Chemistry A, 3(13), 6962-6969 (2015)
Crystal structures, fluorescent and magnetic properties of five new coordination polymers based on biphenyl-3, 4?, 5-tricarboxylic acid
Lu Y, et al.
Journal of Solid State Chemistry, 192, 144-152 (2012)
Synthesis, structure and luminescent property of a new europium-based coordination polymer constructed from biphenyl-3, 4?, 5-tricarboxylic acid.
Yan D and Duan Q
Inorganic Chemistry Communications, 36, 188-191 (2013)

Articles

Metal-organic frameworks offer high surface area materials for alternative energy applications through simple synthetic strategies.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service