Skip to Content
Merck
All Photos(2)

Key Documents

721247

Sigma-Aldrich

2,2′-Diamino-4,4′-stilbenedicarboxylic acid

greener alternative
Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$1,120.00

$1,120.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
$1,120.00

About This Item

Empirical Formula (Hill Notation):
C16H14N2O4
CAS Number:
Molecular Weight:
298.29
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23

$1,120.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

solid

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

200-205 °C

greener alternative category

SMILES string

Nc1cc(ccc1\C=C\c2ccc(cc2N)C(O)=O)C(O)=O

InChI

1S/C16H14N2O4/c17-13-7-11(15(19)20)5-3-9(13)1-2-10-4-6-12(16(21)22)8-14(10)18/h1-8H,17-18H2,(H,19,20)(H,21,22)/b2-1+

InChI key

QEDSDXRFNGAJKN-OWOJBTEDSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

2,2′-Diamino-4,4′-stilbenedicarboxylic acid is an organic linker and an amine-functionalized dicarboxylic acid. It can be used in the formation of metal-organic frameworks (MOFs) for the storage of hydrogen and the reduction of CO2.[1][2][3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enhancement of visible-light-driven CO 2 reduction performance using an amine-functionalized zirconium metal-organic framework
Sun M, et al.
Dalton Transactions, 47(3), 909-915 (2018)
Characteristics of MOF, MWCNT and graphene containing materials for hydrogen storage: A review
Gangu KK, et al.
Chemosphere, 30, 132-144 (2019)
Mixed-linker approach in designing porous zirconium-based metal-organic frameworks with high hydrogen storage capacity
Naeem A, et al.
Chemical Communications (Cambridge, England), 52(50), 7826-7829 (2016)

Articles

Metal-organic frameworks offer high surface area materials for alternative energy applications through simple synthetic strategies.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service