Skip to Content
Merck
All Photos(1)

Key Documents

711225

Sigma-Aldrich

2′,3′-Dihydroxy-4′-methoxyacetophenone hydrate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H10O4 · xH2O
Molecular Weight:
182.17 (anhydrous basis)
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

97%

form

solid

mp

125-130 °C

functional group

ketone

SMILES string

O.COc1ccc(C(C)=O)c(O)c1O

InChI

1S/C9H10O4.H2O/c1-5(10)6-3-4-7(13-2)9(12)8(6)11;/h3-4,11-12H,1-2H3;1H2

InChI key

LLNNBEWBUXDRMW-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service