Skip to Content
Merck
All Photos(1)

Key Documents

710652

Sigma-Aldrich

Sodium bis(2-ethylhexyl-d17) sulfosuccinate

98 atom % D, 96% (CP)

Synonym(s):

Docusate sodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$529.00
100 MG
$2,120.00

$529.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
10 MG
$529.00
100 MG
$2,120.00

About This Item

Empirical Formula (Hill Notation):
C20D34H3NaO7S
CAS Number:
Molecular Weight:
478.77
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.12

$529.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

isotopic purity

98 atom % D

Quality Level

Assay

96% (CP)

form

solid

mol wt

478.09 by atom % calculation

technique(s)

bio NMR: suitable

mass shift

M+34

SMILES string

[Na+].[2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])(C([2H])([2H])OC(=O)CC(C(=O)OC([2H])([2H])C([2H])(C([2H])([2H])C([2H])([2H])[2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H])S([O-])(=O)=O)C([2H])([2H])C([2H])([2H])[2H]

InChI

1S/C20H38O7S.Na/c1-5-9-11-16(7-3)14-26-19(21)13-18(28(23,24)25)20(22)27-15-17(8-4)12-10-6-2;/h16-18H,5-15H2,1-4H3,(H,23,24,25);/q;+1/p-1/i1D3,2D3,3D3,4D3,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,14D2,15D2,16D,17D;

InChI key

APSBXTVYXVQYAB-MTXGETQSSA-M

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Pamela Brunswick et al.
Journal of environmental science and health. Part A, Toxic/hazardous substances & environmental engineering, 55(7), 788-799 (2020-04-01)
Observations made for the analysis of the oil spill dispersant tracer dioctyl sulfosuccinate (DOSS) during LC50 toxicity testing, highlighted a stability issue for this tracer compound in seawater. A liquid chromatography high-resolution quadrupole time-of-flight mass spectrometry (LC/QToF) was used to
Pamela Brunswick et al.
Journal of chromatography. A, 1598, 113-121 (2019-04-17)
A high resolution mass spectrometry method was developed for the environmental impact monitoring of oil spill dispersants. Previously reported instability of dioctyl sulfosuccinate (DOSS) dispersant tracer was addressed by the new procedure. The method monitors both DOSS and its degradation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service