Skip to Content
Merck
All Photos(2)

Key Documents

710318

Sigma-Aldrich

Cyclobutylzinc bromide

0.5 M in THF

Synonym(s):

Cyclobutylzinc(II) bromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H7BrZn
Molecular Weight:
200.39
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22
Product 710318 is currently not available for purchase. Contact Technical Service

form

liquid

reaction suitability

reaction type: C-C Bond Formation

concentration

0.5 M in THF

density

0.984 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

Br[Zn]C1CCC1

InChI

1S/C4H7.BrH.Zn/c1-2-4-3-1;;/h1H,2-4H2;1H;/q;;+1/p-1

InChI key

BBKCOQYMKGEAAJ-UHFFFAOYSA-M

Legal Information

Product of Rieke Metals, Inc.
Rieke is a registered trademark of Rieke Metals, Inc.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3 - Water-react 2

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service