Skip to Content
Merck
All Photos(1)

Key Documents

498041

Sigma-Aldrich

Cyclopentylzinc bromide solution

0.5 M in THF

Synonym(s):

Bromo(cyclopentyl)zinc, Bromocyclopentylzinc

Sign Into View Organizational & Contract Pricing

Select a Size

50 ML
$433.00

$433.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 ML
$433.00

About This Item

Linear Formula:
C5H9ZnBr
CAS Number:
Molecular Weight:
214.42
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

$433.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

concentration

0.5 M in THF

density

0.955 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

Br[Zn]C1CCCC1

InChI

1S/C5H9.BrH.Zn/c1-2-4-5-3-1;;/h1H,2-5H2;1H;/q;;+1/p-1

InChI key

GOPYCBPMCCJYSN-UHFFFAOYSA-M

Application

Cyclopentylzinc bromide is an organozinc reagent that can be used in:
  • The synthesis of polyfunctional indoles by reacting with various aryldiazonium salts via Fischer indole synthesis.[1]
  • Negishi cross-coupling reactions with various haloarenes in presence of Pd-complex catalyst.[2]
  • The preparation of aryl alkyl ketones by alkylation of aryl N-methyl-N-tosyl or N-benzyl-N-(tert-butoxycarbonyl) amide derivatives in the presence of Ni catalyst and imidazolidinylidene ligand.[3]

Legal Information

Product of Rieke Metals, Inc.
Rieke is a registered trademark of Rieke Metals, Inc.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3 - Water-react 2

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Efficient preparation of polyfunctional indoles via a zinc organometallic variation of the Fischer indole synthesis
Zhang Z-G, et al.
Synthesis, 2011(01), 23-29 (2011)
Nickel-catalyzed alkylation of amide derivatives
Simmons BJ, et al.
ACS Catalysis, 6(5), 3176-3179 (2016)
Mild Negishi cross-coupling reactions catalyzed by acenaphthoimidazolylidene palladium complexes at low catalyst loadings
Liu Z, et al.
The Journal of Organic Chemistry, 78(15), 7436-7444 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service