Skip to Content
Merck
All Photos(2)

Key Documents

709832

Sigma-Aldrich

3-Fluoro-2-formylphenylboronic acid

≥95%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$75.50

$75.50

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$75.50

About This Item

Empirical Formula (Hill Notation):
C7H6BFO3
CAS Number:
Molecular Weight:
167.93
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

$75.50

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥95%

form

powder

mp

123-127 °C

functional group

aldehyde
fluoro

storage temp.

2-8°C

SMILES string

OB(O)c1cccc(F)c1C=O

InChI

1S/C7H6BFO3/c9-7-3-1-2-6(8(11)12)5(7)4-10/h1-4,11-12H

InChI key

GVHWLCYABLWGIR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

May contain varying amounts of anhydride

Application

  • Undergoes unprecedented tautomeric rearrangement in solution

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service