Skip to Content
Merck
All Photos(2)

Key Documents

657344

Sigma-Aldrich

2-Fluoro-4-formylphenylboronic acid

≥95.0%

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$388.00

$388.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$388.00

About This Item

Empirical Formula (Hill Notation):
C7H6BFO3
CAS Number:
Molecular Weight:
167.93
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

$388.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥95.0%

form

solid

mp

161-178 °C

functional group

aldehyde
fluoro

SMILES string

OB(O)c1ccc(C=O)cc1F

InChI

1S/C7H6BFO3/c9-7-3-5(4-10)1-2-6(7)8(11)12/h1-4,11-12H

InChI key

OAEJODVYVOAOPH-UHFFFAOYSA-N

Other Notes

Contains varying amounts of anhydride

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service