Skip to Content
Merck
All Photos(1)

Key Documents

709239

Sigma-Aldrich

Bismuth tungsten oxide

powder, −200 mesh, 99.5% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Bi2(WO4)3
CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:

Assay

99.5% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: bismuth

reagent type: catalyst
core: tungsten

particle size

−200 mesh

mp

>300 °C

SMILES string

[Bi+3].[Bi+3].[O-][W]([O-])(=O)=O.[O-][W]([O-])(=O)=O.[O-][W]([O-])(=O)=O

InChI

1S/2Bi.12O.3W/q2*+3;;;;;;;6*-1;;;

InChI key

DWNBMQNJXHTBPG-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service