Skip to Content
Merck
All Photos(1)

Key Documents

570680

Sigma-Aldrich

Benzene-D6

anhydrous, ≥99.6 atom % D

Synonym(s):

Hexadeuterobenzene

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
$555.00

$555.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 G
$555.00

About This Item

Empirical Formula (Hill Notation):
C6D6
CAS Number:
Molecular Weight:
84.15
Beilstein:
1905426
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.11

$555.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

anhydrous

Quality Level

isotopic purity

≥99.6 atom % D

Assay

≥99.0%

form

liquid

expl. lim.

0.34-6.3 % (lit.)

technique(s)

NMR: suitable

impurities

<10 ppm water

refractive index

n20/D 1.497 (lit.)

bp

79.1 °C/760 mmHg (lit.)

mp

6.8 °C (lit.)

density

0.950 g/mL at 25 °C (lit.)

mass shift

M+6

SMILES string

[2H]c1c([2H])c([2H])c([2H])c([2H])c1[2H]

InChI

1S/C6H6/c1-2-4-6-5-3-1/h1-6H/i1D,2D,3D,4D,5D,6D

InChI key

UHOVQNZJYSORNB-MZWXYZOWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Benzene-d6 (C6D6) is deuterated benzene. It is an anhydrous solvent, which minimizes any interference from water peaks. It is useful in performing critical air- and moisture-sensitive NMR analysis. Its Soret coefficient S(T) has been measured by transient holographic grating technique.[1] Its synthesis,13C NMR, IR and MS spectra have been reported.[2][3]

Application

Benzene-d6 (C6D6) may be used in the following:
  • Synthesis of isotopically labeled samples.[4]
  • As a 1H and 13C-NMR solvent.[5][6]
  • In the internal calibration of benzene stock solution.[7]

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Carc. 1A - Eye Irrit. 2 - Flam. Liq. 2 - Muta. 1B - Skin Irrit. 2 - STOT RE 1

Target Organs

Blood

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

12.2 °F - closed cup

Flash Point(C)

-11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Synthesis and Spectral Analysis of Benzene-d6.
Guo YZ, et al.
Advanced Materials Research, 1061-1062, 301-306 (2014)
Surprising Titanium complexes bearing η2-pyrazolato ligands: synthesis, structure, and molecular orbital studies.
Guzei IA, et al.
Journal of the American Chemical Society, 119(14), 3387-3388 (1997)
Structure-property relationships in nematic gold nanoparticles.
Cseh L and Mehl GH.
Journal of Materials Chemistry, 17(4), 311-315 (2007)
Biomonitoring of benzene and toluene in human blood by headspace-solid-phase microextraction.
Schimming E, et al.
Fresenius Journal of Analytical Chemistry, 363(1), 88-91 (1999)

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service