Skip to Content
Merck
All Photos(2)

Documents

706884

Sigma-Aldrich

1,3,5-Tris(4′-carboxy[1,1′-biphenyl]-4-yl)benzene

greener alternative

95%

Synonym(s):

H3BBC, H3TCBPB, TCBTB

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C45H30O6
CAS Number:
Molecular Weight:
666.72
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23

Assay

95%

form

powder

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

>300 °C

greener alternative category

SMILES string

OC(=O)c1ccc(cc1)-c2ccc(cc2)-c3cc(cc(c3)-c4ccc(cc4)-c5ccc(cc5)C(O)=O)-c6ccc(cc6)-c7ccc(cc7)C(O)=O

InChI

1S/C45H30O6/c46-43(47)37-19-13-31(14-20-37)28-1-7-34(8-2-28)40-25-41(35-9-3-29(4-10-35)32-15-21-38(22-16-32)44(48)49)27-42(26-40)36-11-5-30(6-12-36)33-17-23-39(24-18-33)45(50)51/h1-27H,(H,46,47)(H,48,49)(H,50,51)

InChI key

PEQRGMPXYDIZSX-UHFFFAOYSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

1,3,5-Tris(4′-carboxy[1,1′-biphenyl]-4-yl)benzene is used as a linker molecule for the synthesis of high performance metal-organic framework (MOFs).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Selected Applications of Metal-Organic Frameworks in Sustainable Energy Technologies?.
Caskey SR and Matzger AJ.
Material Matters, 4(4), 111-111 (2009)

Articles

Metal-organic frameworks offer high surface area materials for alternative energy applications through simple synthetic strategies.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service