Skip to Content
Merck
All Photos(1)

Key Documents

259810

Sigma-Aldrich

2,4,6-Triphenyl-1,3,5-triazine

98%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$162.00

$162.00


Estimated to ship onMarch 30, 2025


Request a Bulk Order

Select a Size

Change View
5 G
$162.00

About This Item

Empirical Formula (Hill Notation):
C21H15N3
CAS Number:
Molecular Weight:
309.36
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
98%

$162.00


Estimated to ship onMarch 30, 2025


Request a Bulk Order

Quality Level

Assay

98%

mp

232-235 °C (lit.)

functional group

phenyl

SMILES string

c1ccc(cc1)-c2nc(nc(n2)-c3ccccc3)-c4ccccc4

InChI

1S/C21H15N3/c1-4-10-16(11-5-1)19-22-20(17-12-6-2-7-13-17)24-21(23-19)18-14-8-3-9-15-18/h1-15H

InChI key

HBQUOLGAXBYZGR-UHFFFAOYSA-N

General description

Hole blocking capability of 2,4,6-triphenyl-1,3,5-triazine as a non-transporting filler material has been investigated[1].

Application

2,4,6-Triphenyl-1,3,5-triazine has been used as a hole-blocking material for electroluminescent devices (ELD)[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Hole-blocking characteristics of 2, 4, 6-triphenyl-1, 3, 5-triazine as a non-transporting filler material.
Myung Shin D, et al.
Thin Solid Films, 363(1), 252-254 (2000)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service