Skip to Content
Merck
All Photos(1)

Key Documents

702641

Sigma-Aldrich

Thieno[2,3-b]thiophene

95%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$609.00

$609.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Select a Size

Change View
5 G
$609.00

About This Item

Empirical Formula (Hill Notation):
C6H4S2
CAS Number:
Molecular Weight:
140.23
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

$609.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Assay

95%

form

liquid

refractive index

n20/D 1.668

density

1.326 g/cm3 at 25 °C

SMILES string

c1cc2ccsc2s1

InChI

1S/C6H4S2/c1-3-7-6-5(1)2-4-8-6/h1-4H

InChI key

YHBTXTFFTYXOFV-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

199.9 °F

Flash Point(C)

93.3 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service