Skip to Content
Merck
All Photos(1)

Key Documents

701998

Sigma-Aldrich

[1,1′-Bis(di-cyclohexylphosphino)ferrocene]dichloropalladium(II)

98%

Synonym(s):

PdCl2(dcypf)

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$74.10
1 G
$282.00

$74.10

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
$74.10
1 G
$282.00

About This Item

Empirical Formula (Hill Notation):
C34H52Cl2FeP2Pd
CAS Number:
Molecular Weight:
755.90
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

$74.10

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

powder

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Cross Couplings
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

mp

294-300 °C

storage temp.

−20°C

SMILES string

[Fe].Cl[Pd]Cl.[CH]1[CH][CH][C]([CH]1)P(C2CCCCC2)C3CCCCC3.[CH]4[CH][CH][C]([CH]4)P(C5CCCCC5)C6CCCCC6

InChI

1S/2C17H26P.2ClH.Fe.Pd/c2*1-3-9-15(10-4-1)18(17-13-7-8-14-17)16-11-5-2-6-12-16;;;;/h2*7-8,13-16H,1-6,9-12H2;2*1H;;/q;;;;;+2/p-2

InChI key

HRAMBTUEZQOQRK-UHFFFAOYSA-L

Application

1,1′-Bis(di-cyclohexylphosphino)ferrocene]dichloropalladium(II) or (Pd(dcpf)Cl2) can be used as a catalyst in the synthesis of:
  • α, β-unsaturated amides by hydroaminocarbonylation of alkynes with tertiary amines.[1]
  • Quinazoline derivatives via Suzuki-Miyaura coupling reaction between of quinazolines containing unprotected NH2 group and arylboronic acids.[2]
  • α-aryl carbonyl compounds by α-arylation of ketones with aryl chlorides and aryl bromides.[3]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Suzuki-Miyaura coupling of quinazolines containing an unprotected NH2 group: Synthesis and biological testing of quinazoline derivatives
Pulipati Y, et al.
Synthetic Communications, 47(12), 1142-1150 (2017)
A highly practical and general route for α-arylations of ketones using Bis-phosphinoferrocene-based palladium catalysts
Grasa GA and Colacot TJ
Organic Process Research & Development, 12(3), 522-529 (2008)
Palladium-catalyzed hydroaminocarbonylation of alkynes with tertiary amines via C-N bond cleavage
Gao, Bao and Huang, Hanmin
Organic Letters, 19(22), 6260-6263 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service