Skip to Content
Merck
All Photos(2)

Key Documents

698865

Sigma-Aldrich

Sodium aluminum hydride solution

1 M in THF

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaAlH4
CAS Number:
Molecular Weight:
54.00
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:

form

liquid

reaction suitability

reagent type: reductant

concentration

1 M in THF

density

0.905 g/mL at 25 °C

SMILES string

[Na+].[AlH4-]

InChI

1S/Al.Na.4H/q-1;+1;;;;

InChI key

MNYCIYRTKITUKR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Reaction kinetics of sodium aluminum hydrides may be improved by mechanical grinding or chemical modification. This may result in achieving reversible hydrogen storage at lower temperatures (80-140oC). It may behave as a reducing agent. Reaction kinetics of graphite and Ti-doped NaAlH4 was investigated. Presence of Ti doping enhances the kinetics of absorption and desorption. TiO2, CeO2, La2O3, Pr2O3, Nd2O3, Sm2O3, Eu2O3 and Gd2O3 metal oxides were studied for their catalytic behavior in improving the storage kinetics of NAlH4.

Application

NaAlH4 may be used in the deoxygenative dimerization of carbonyls and alcohols and in hydroalumination of alkenes and alkynes.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3 - Water-react 2

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 1

Flash Point(F)

-7.6 °F - closed cup

Flash Point(C)

-22 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Titanium?halide catalyst-precursors in sodium aluminum hydrides.
Majzoub EH and Gross KJ
J. Alloy Compounds, 356, 363-367 (2003)
Effect of graphite as a co-dopant on the dehydrogenation and hydrogenation kinetics of Ti-doped sodium aluminum hydride
Wang J,et al.
J. Alloy Compounds, 395(1-2), 252-262 (2005)
Studies on metal oxide nanoparticles catalyzed sodium aluminum hydride.
Pukazhselvan D
Energy, 35(12), 5037-5042 (2010)
Gugelchuk M, et al. et al.
e-EROS Encyclopedia of Reagents for Organic Synthesis. (2009)
Sodium alanates for reversible hydrogen storage,
Zaluska A, et al.
J. Alloy Compounds, 298(1-2), 125-134 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service