Skip to Content
Merck
All Photos(1)

Key Documents

C84801

Sigma-Aldrich

Coronene

97%

Synonym(s):

Superbenzene, [6]Circulene

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$138.00
500 MG
$559.00

$138.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$138.00
500 MG
$559.00

About This Item

Empirical Formula (Hill Notation):
C24H12
CAS Number:
Molecular Weight:
300.35
Beilstein:
658468
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$138.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

powder

bp

525 °C (lit.)

mp

428 °C (lit.)

SMILES string

c1cc2ccc3ccc4ccc5ccc6ccc1c7c2c3c4c5c67

InChI

1S/C24H12/c1-2-14-5-6-16-9-11-18-12-10-17-8-7-15-4-3-13(1)19-20(14)22(16)24(18)23(17)21(15)19/h1-12H

InChI key

VPUGDVKSAQVFFS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Coronene can be used
  • To synthesize active materials for organic photovoltaic and optoelectronic applications.[1][2]
  • As ′nucleation seed′ to synthesize graphene by vapor deposition at low temperatures.[3]
  • To build novel polycyclic aromatic molecular frameworks and architectures.[4][5][6]

Incorporation of coronene as ′Carbon Dot′ for bioimaging applications has been reported.[7]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Persulfurated Coronene: A New Generation of ?Sulflower?.
Dong R, et al.
Journal of the American Chemical Society, 139(6), 2168-2171 (2017)
Luminescent Carbon Dot Mimics Assembled on DNA.
Chan KM et al.
Journal of the American Chemical Society, 139(37), 13147-13155 (2017)
Continuous graphene films synthesized at low temperatures by introducing coronene as nucleation seeds.
Wu T, et al.
Nanoscale, 5(12), 5456-5461 (2013)
Porphyrins fused with unactivated polycyclic aromatic hydrocarbons.
Diev VV, et al.
The Journal of Organic Chemistry, 77(1), 143-159 (2011)
Perchlorocoronene: a novel host precursor.
Baird T, et al.
Journal of the Chemical Society. Chemical Communications, (22), 1471-1473 (1988)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service