Skip to Content
Merck
All Photos(1)

Key Documents

693286

Sigma-Aldrich

(S)-Ru(OAc)2(DM-BINAP)

Synonym(s):

Diacetato[(S)-(−)-2,2′-bis[di(3,5-xylyl)phosphino]-1,1′−binaphthyl]ruthenium(II)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C56H54O4P2Ru
CAS Number:
Molecular Weight:
954.04
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

form

powder

functional group

phosphine

storage temp.

2-8°C

SMILES string

CC(=O)O[Ru]OC(C)=O.Cc1cc(C)cc(c1)P(c2cc(C)cc(C)c2)c3ccc4ccccc4c3-c5c(ccc6ccccc56)P(c7cc(C)cc(C)c7)c8cc(C)cc(C)c8

InChI

1S/C52H48P2.2C2H4O2.Ru/c1-33-21-34(2)26-43(25-33)53(44-27-35(3)22-36(4)28-44)49-19-17-41-13-9-11-15-47(41)51(49)52-48-16-12-10-14-42(48)18-20-50(52)54(45-29-37(5)23-38(6)30-45)46-31-39(7)24-40(8)32-46;2*1-2(3)4;/h9-32H,1-8H3;2*1H3,(H,3,4);/q;;;+2/p-2

InChI key

IFUMNSKQNPECEY-UHFFFAOYSA-L

Legal Information

Sold in collaboration with Takasago for research purposes only.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Hydrogenation, Asymmetric Catalysis, Binap, SEGPHOS®, Aldol reaction, Alkenylation, Arylation, Mannich reaction, Fluorination, Michael addition, Hydrosilylation, Cycloaddition, Takasago

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service