Skip to Content
Merck
All Photos(4)

Key Documents

698717

Sigma-Aldrich

Polyoxyethanyl-α-tocopheryl sebacate

greener alternative

15 wt. % in H2O

Synonym(s):

PTS

Sign Into View Organizational & Contract Pricing

Select a Size

10 ML
$128.00
100 ML
$590.00

$128.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 ML
$128.00
100 ML
$590.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.22

$128.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

liquid

mol wt

~1,200

reaction suitability

reaction type: C-C Bond Formation

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

15 wt. % in H2O

greener alternative category

General description

Learn More at the Professor and Product Portal of Professor Bruce Lipshutz.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced to increase the catalytic efficiency and acts as an environmentally benign and sustainable amphiphile. Click here for more information.

Application

PTS: Amphiphile for Organic Synthesis in Water

Uses:
  • Provides an aqueous micellar environment for transition metal-catalyzed Heck coupling, cross metathesis and ring closing metathesis reactions
  • Surfactant for catalytic asymmetric Baeyer-Villiger oxidation in water using PtII catalysts and hydrogen peroxide
  • Orally active CoQ10 carrier for neuroprotection after stroke or cardiac arrest

Legal Information

Product of Zymes LLC sold under exclusive license to US and worldwide patents.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Micellular catalysis has provided the ability to carry out several commonly used transformations used in the synthetic community to be carried out in water.

Surfactants for efficient reactions in water as a green chemistry alternative.

The Baeyer-Villiger oxidation is the oxidative cleavage of a carbon-carbon bond adjacent to a carbonyl, which converts the ketones to esters and the cyclic ketones to lactones.

Related Content

Designer surfactants enable diverse transformations like Suzuki-Miyaura, Olefin Metathesis, and 1,4-Addition to Enones in water by Prof. Bruce Lipshutz.

Questions

1–2 of 2 Questions  
  1. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  2. What can Product 698717, Polyoxyethanyl-α-tocopheryl sebacate, be used for?

    1 answer
    1. PTS is a non-ionic amphiphile that can be used to "solubilize" organic molecules in aqueous media. The PTS molecule forms micelles around the lipophilic compounds, allowing them to move into the aqueous phase for effective cross-coupling and metathesis reactions without the need of a co-solvent.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service