Skip to Content
Merck
All Photos(2)

Key Documents

692786

Sigma-Aldrich

(S,S)-DACH-naphthyl Trost Ligand

95%

Synonym(s):

(1S,2S)-(–)-1,2-Diaminocyclohexane-N,N′-bis(2-diphenylphosphino-1-naphthoyl)

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$91.10
1 G
$347.00

$91.10

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
250 MG
$91.10
1 G
$347.00

About This Item

Empirical Formula (Hill Notation):
C52H44N2O2P2
CAS Number:
Molecular Weight:
790.87
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

$91.10

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Assay

95%

form

solid

optical activity

[α]20/D -66.0°, c = 1 in methanol

mp

232-237 °C

functional group

amide
phosphine

SMILES string

O=C(N[C@H]1CCCC[C@@H]1NC(=O)c2c(ccc3ccccc23)P(c4ccccc4)c5ccccc5)c6c(ccc7ccccc67)P(c8ccccc8)c9ccccc9

InChI

1S/C52H44N2O2P2/c55-51(49-43-29-15-13-19-37(43)33-35-47(49)57(39-21-5-1-6-22-39)40-23-7-2-8-24-40)53-45-31-17-18-32-46(45)54-52(56)50-44-30-16-14-20-38(44)34-36-48(50)58(41-25-9-3-10-26-41)42-27-11-4-12-28-42/h1-16,19-30,33-36,45-46H,17-18,31-32H2,(H,53,55)(H,54,56)/t45-,46-/m0/s1

InChI key

VXFKMKXTPXVEMU-ZYBCLOSLSA-N

Application

Asymmetric allylic alkylation ligand.[1][2][3][4]

Legal Information

Sold in collaboration with Dr. Reddy′s Laboratories for research purposes only.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Trost, B.M. et al.
Journal of the American Chemical Society, 124, 11616-11616 (2004)
Asymmetric Transition Metal-Catalyzed Allylic Alkylations.
Barry M. Trost et al.
Chemical reviews, 96(1), 395-422 (1996-02-01)
Trost, B.M. et al.
Aldrichimica Acta, 40, 59-59 (2007)
Asymmetric transition-metal-catalyzed allylic alkylations: applications in total synthesis.
Barry M Trost et al.
Chemical reviews, 103(8), 2921-2944 (2003-08-14)

Articles

Trost group pioneers C-2 symmetric ligands for rapid chiral product synthesis in asymmetric catalytic reactions.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service