Skip to Content
Merck
All Photos(1)

Key Documents

692794

Sigma-Aldrich

(S,S)-DACH-phenyl Trost ligand

95%

Synonym(s):

(1S,2S)-(–)-1,2-Diaminocyclohexane-N,N′-bis(2-diphenylphosphinobenzoyl)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H40N2O2P2
CAS Number:
Molecular Weight:
690.75
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.06
Pricing and availability is not currently available.

Assay

95%

form

solid

optical activity

[α]20/D -134°, c = 1 in methanol

mp

136-142 °C

functional group

amide
phosphine

SMILES string

O=C(N[C@H]1CCCC[C@@H]1NC(=O)c2ccccc2P(c3ccccc3)c4ccccc4)c5ccccc5P(c6ccccc6)c7ccccc7

InChI

1S/C44H40N2O2P2/c47-43(37-27-13-17-31-41(37)49(33-19-5-1-6-20-33)34-21-7-2-8-22-34)45-39-29-15-16-30-40(39)46-44(48)38-28-14-18-32-42(38)50(35-23-9-3-10-24-35)36-25-11-4-12-26-36/h1-14,17-28,31-32,39-40H,15-16,29-30H2,(H,45,47)(H,46,48)/t39-,40-/m0/s1

InChI key

AXMSEDAJMGFTLR-ZAQUEYBZSA-N

Application

Asymmetric allylic alkylation ligand.[1][2][3][4]

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

Sold in collaboration with Dr. Reddy′s Laboratories for research purposes only.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aldrichimica Acta, 40, 59-59 (2007)
Trost, B.M. et al.
Aldrichimica Acta, 40, 59-59 (2007)
Asymmetric Transition Metal-Catalyzed Allylic Alkylations.
Barry M. Trost et al.
Chemical reviews, 96(1), 395-422 (1996-02-01)
Trost, B.M. et al.
Journal of the American Chemical Society, 124, 11616-11616 (2004)
Asymmetric transition-metal-catalyzed allylic alkylations: applications in total synthesis.
Barry M Trost et al.
Chemical reviews, 103(8), 2921-2944 (2003-08-14)

Articles

Trost group pioneers C-2 symmetric ligands for rapid chiral product synthesis in asymmetric catalytic reactions.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service