691372
1,3-Dihydroxy-2-methylimidazolium bis(trifluoromethylsulfonyl)imide
98%
Synonym(s):
(OH)2MeIm-NTF2
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C6H7F6N3O6S2
CAS Number:
Molecular Weight:
395.26
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
≥98% (13C-NMR)
≥98% (19F-NMR)
≥98% (1H-NMR)
98%
form
liquid
mp
72 °C
suitability
complies for IR spectroscopy
functional group
fluoro
SMILES string
Cc1n(O)cc[n+]1O.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F
InChI
1S/C4H7N2O2.C2F6NO4S2/c1-4-5(7)2-3-6(4)8;3-1(4,5)14(10,11)9-15(12,13)2(6,7)8/h2-3,7-8H,1H3;/q+1;-1
InChI key
VQKFGWWOHFMUPX-UHFFFAOYSA-N
Application
Ionic liquid and NHC precatalyst
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
N,N-Di(alkoxy)imidazolium salts: new patent-free ionic liquids and NHC-precatalysts
Laus, G., et al.
Zeitschrift fur Naturforschung, B: Chemical Sciences, 62, 295-308 (2007)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service