Skip to Content
Merck
All Photos(1)

Documents

375241

Sigma-Aldrich

Propyl vinyl ether

99%

Synonym(s):

1-(Ethenyloxy)propane, 1-(Vinyloxy)propane, n -Propyl vinyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2OCH=CH2
CAS Number:
Molecular Weight:
86.13
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

form

liquid

autoignition temp.

365 °F

contains

0.1% potassium hydroxide as inhibitor

refractive index

n20/D 1.391 (lit.)

bp

65 °C (lit.)

density

0.768 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCCOC=C

InChI

1S/C5H10O/c1-3-5-6-4-2/h4H,2-3,5H2,1H3

InChI key

OVGRCEFMXPHEBL-UHFFFAOYSA-N

Related Categories

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-14.8 °F - closed cup

Flash Point(C)

-26 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

O Schalk et al.
The journal of physical chemistry. A, 119(45), 11105-11112 (2015-10-23)
A series of different alkyl vinyl ethers is investigated to decipher the possible reaction channels upon photoexcitation to the π3s-Rydberg and the ππ*-valence state at 200 nm using time-resolved photoelectron spectroscopy and on-the-fly time-dependent density functional theory dynamics simulations. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service