Skip to Content
Merck
All Photos(1)

Documents

689718

Sigma-Aldrich

Benzyldimethylhexylammonium chloride

≥96.0% (AT)

Synonym(s):

Benzylhexyldimethylammonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H26ClN
CAS Number:
Molecular Weight:
255.83
Beilstein:
4045485
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

≥96.0% (AT)

form

powder

SMILES string

[Cl-].CCCCCC[N+](C)(C)Cc1ccccc1

InChI

1S/C15H26N.ClH/c1-4-5-6-10-13-16(2,3)14-15-11-8-7-9-12-15;/h7-9,11-12H,4-6,10,13-14H2,1-3H3;1H/q+1;/p-1

InChI key

BNDXNEVHWSOJGV-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service