Skip to Content
Merck
All Photos(1)

Documents

689599

Sigma-Aldrich

Benzyldimethyloctylammonium chloride

≥96.0% (AT)

Synonym(s):

N,N-dimethyl-N-octyl-benzenemethanaminium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H30ClN
CAS Number:
Molecular Weight:
283.88
Beilstein:
3771220
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

≥96.0% (AT)

form

powder

SMILES string

[Cl-].CCCCCCCC[N+](C)(C)Cc1ccccc1

InChI

1S/C17H30N.ClH/c1-4-5-6-7-8-12-15-18(2,3)16-17-13-10-9-11-14-17;/h9-11,13-14H,4-8,12,15-16H2,1-3H3;1H/q+1;/p-1

InChI key

PXFDQFDPXWHEEP-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Ines Mulder et al.
Environmental science and pollution research international, 27(36), 45311-45323 (2020-08-14)
Quaternary alkyl ammonium compounds (QAACs) are produced in large quantities for use as surfactants and disinfectants and also found in soils, sediments, and surface waters, where they are potentially involved in the selection of antibiotic resistance genes. Micelle formation influences
Mlamuli Ndabambi et al.
Chemosphere, 247, 125924-125924 (2020-01-25)
Due to their use in various domestic and industrial formulations, benzalkonium compounds have been isolated in many environmental matrices. Sorption to soil components has been shown to play a key role in their environmental fate. Whereas sorption of benzalkonium compounds
Kahina Slimani et al.
Journal of chromatography. A, 1517, 86-96 (2017-08-31)
Quaternary ammonium compounds (QACs) are both cationic surfactants and biocidal substances widely used as disinfectants in the food industry. A sensitive and reliable method for the analysis of benzalkonium chlorides (BACs) and dialkyldimethylammonium chlorides (DDACs) has been developed that enables

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service